missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FoxJ1 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems IC3619U-100UG
This item is not returnable.
View return policy
Description
FoxJ1/HFH4 Monoclonal specifically detects FoxJ1/HFH4 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| FoxJ1/HFH4 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q92949.3 | |
| FOXJ1 | |
| E. coli-derived recombinant human FoxJ1 Lys306-Leu421 Accession # Q92949.3 | |
| RUO | |
| 2302 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 407003 | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| FKHL13, FKHL13forkhead transcription factor HFH-4, forkhead box J1, forkhead-like 13, Forkhead-related protein FKHL13, Hepatocyte nuclear factor 3 forkhead homolog 4, HFH-4, HFH4fork head homologue 4, HFH-4forkhead box protein J1, MGC35202 | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects human FoxJ1 in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction