missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FoxP1 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC45341R-100UG
This item is not returnable.
View return policy
Description
FoxP1 Monoclonal specifically detects FoxP1 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| FoxP1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q9H334 | |
| FOXP1 | |
| E. coli-derived recombinant human FoxP1 Lys548-Glu677 Accession # Q9H334 | |
| RUO | |
| 27086 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 837016 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| FLJ23741, fork head-related protein like B, forkhead box P1, forkhead box protein P1, glutamine-rich factor 1, hFKH1B, HSPC215, MGC12942, MGC88572, MGC99551, QRF1, QRF112CC4 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human FoxP1 in direct ELISAs and Western blots. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction