missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FoxP4 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF6854-SP
This item is not returnable.
View return policy
Description
FoxP4 Polyclonal specifically detects FoxP4 in Human samples. It is validated for Western Blot.
Specifications
| FoxP4 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FKHLA, FLJ40908, FLJ44184, fork head-related protein like A, Fork head-related protein-like A, forkhead box P4, forkhead box protein P4, hFKHLA, winged-helix repressor FOXP4 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 116113 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1:1000 dilution | |
| Q8IVH2 | |
| FOXP4 | |
| E. coli-derived recombinant human FoxP4 Pro549-Ser680 Accession # Q8IVH2 | |
| 25 μg | |
| Primary | |
| Detects human FoxP4 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) FoxP1, rhFoxP2, rhFoxP3, and rhCTBP1 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction