missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FRA-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB4935-SP
This item is not returnable.
View return policy
Description
FRA-1/FOSL1 Monoclonal specifically detects FRA-1/FOSL1 in Human samples. It is validated for Western Blot.
Specifications
| FRA-1/FOSL1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P15407 | |
| FOSL1 | |
| E. coli-derived recombinant human FRA-1, Gly146-Ala247, Accession # P15407 | |
| 25 μg | |
| Primary | |
| Detects human FRA-1 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human FRA-2 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 496106 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FOSL1, FOS-like antigen 1, FOS-like antigen-1, fos-related antigen 1, FRA, FRA1, FRA-1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8061 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction