Learn More
Gibco™ Human Furin Recombinant Protein, PeproTech®
Description
450-47-100UG will be provided as 2 x 50 μg (450-47-50UG). 450-47-1MG will be provided as 20 x 50 μg (450-47-50UG). 450-47-250UG will be provided as 5 x 50 μg (450-47-50UG). 450-47-500UG will be provided as 5 x 100 μg (450-47-50UG). Recombinant Human Furin is a 63.9 kDa protein, corresponding to residues 131 through 715 of the Furin precursor, plus a C-terminal His-tag. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Specifications
Specifications
| Accession Number | P09958 |
| For Use With (Application) | Functional Assay |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 5045 |
| Molecular Weight (g/mol) | 63.9 kDa |
| Name | Human Furin |
| Quantity | 10 μg |
| Source | Insect cells |
| Regulatory Status | RUO |
| Endotoxin Concentration | <1 EU/ μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.