missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human G-CSF Quantikine QuicKit ELISA
Click to view available options
Quantity:
1 Kit
Unit Size:
Pack of 96
Specifications
Specifications
| Assay Sensitivity | 3.57 pg/mL |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA), Heparin Plasma, Serum |
| Specificity | Natural and Recombinant Human G-CSF |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 1440 |
| Measuring Range | 39.10 to 2500 pg/mL |
| Quantity | 1 Kit |
| Storage Requirements | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
| Synonym | C17orf33, chromosome 17 open reading frame 33, colony stimulating factor 3 (granulocyte), CSF3, CSF3OS, Filgrastim, GCSF, G-CSF, GCSFlenograstim, granulocyte colony-stimulating factor, Lenograstim, MGC45931, pluripoietin |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction