Learn More
Human GAD1/GAD67 Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems BAF2086
This item is not returnable.
View return policy
Description
GAD1/GAD67 Polyclonal specifically detects GAD1/GAD67 in Human samples. It is validated for Western Blot.
Specifications
| GAD1/GAD67 | |
| Polyclonal | |
| Biotin | |
| Q99259 | |
| GAD1 | |
| E. coli-derived recombinant human GAD1/GAD67 Ala2-Asn97 Accession # Q99259 | |
| 50 μg | |
| Primary | |
| Detects human GAD1/GAD67 in Western blots. In Western blots, less than 1% cross-reactivity with recombinant human GAD2/GAD65 is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| 67kD), EC 4.1.1, FLJ45882, GAD25, GAD67, glutamate decarboxylase 1 (brain, 67kDa), Glutamate decarboxylase 67 kDa isoform | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2571 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |