Learn More
Human GAD1/GAD67 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody
Brand:  R&D Systems BAF2086
Product Code. 18090912
  • € 498.00 / 100µg
Description
GAD1/GAD67 Polyclonal specifically detects GAD1/GAD67 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GAD1/GAD67
Polyclonal
Biotin
Q99259
GAD1
E. coli-derived recombinant human GAD1/GAD67 Ala2-Asn97 Accession # Q99259
50 μg
Primary
Detects human GAD1/GAD67 in Western blots. In Western blots, less than 1% cross-reactivity with recombinant human GAD2/GAD65 is observed.
Human
IgG
Western Blot
LYOPH
Western Blot 0.1 ug/mL
67kD), EC 4.1.1, FLJ45882, GAD25, GAD67, glutamate decarboxylase 1 (brain, 67kDa), Glutamate decarboxylase 67 kDa isoform
Goat
Affinity Purified
RUO
2571
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.