missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Galanin R1/GALR1 Alexa Fluor™ 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB10353V-100UG
This item is not returnable.
View return policy
Description
Galanin R1/GALR1 Monoclonal specifically detects Galanin R1/GALR1 in Human samples. It is validated for Flow Cytometry.
Specifications
| Galanin R1/GALR1 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| GAL1, galanin receptor 1, galanin receptor type 1, GalaninR1, GALNR1, GALNRGALNR1GAL1-R, GALR1, GALR-1 | |
| NS0 mouse myeloma cell line transfected with human Galanin R1/GALR1, Accession # P47211 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 613207 | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P47211 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2587 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction