Learn More
Human Galanin R2/GALR2 Alexa Fluor 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems FAB6544R-100UG
Product Code. 18740548
  • € 492.00 / 100µg
Description
Galanin R2/GALR2 Monoclonal specifically detects Galanin R2/GALR2 in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Galanin R2/GALR2
Monoclonal
Alexa Fluor 647
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
GAL2R, GAL2-R, galanin receptor 2, galanin receptor type 2, GalaninR2, GALNR2, GALNR2MGC125984, GALR2, GALR-2, MGC125983
NS0 mouse myeloma cell line transfected with human Galanin R2, Accession # O43603
100 μg
8811
Human
Supernatant
Flow Cytometry
616520
Flow Cytometry 0.25-1 μg/106 cells
O43603
Mouse
Protein A or G purified from hybridoma culture supernatant
Primary
Detects human GAL-R2 in direct ELISAs.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
IgG2b