missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Galectin 1 (aa 12-127) Control Fragment Recombinant Protein

Product Code. 30200831
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30200831

Brand: Invitrogen™ RP104541

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (87%), Rat (87%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Galectin-1 belongs to a large family of carbohydrate-binding proteins called lectins. Galectin-1can be either monomeric or homodimeric and is found in a wide variety of cells and tissue types. Galectin-1 can control cell growth, proliferation, induce apoptosis of activated T cells while it can also modulate cytokine secretion or inhibit pro-inflammatory cytokine production. Galectin-1 plays an important role in acute and chronic inflammation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P09382
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 3956
Name Human Galectin 1 (aa 12-127) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 14 kDa laminin-binding protein; 14 kDa lectin; AA410090; beta-galactoside binding protein; beta-galactoside-binding lectin; beta-galactoside-binding lectin L-14-I; beta-galactoside-binding protein 14 kDa; DKFZp686E23103; GAL1; GAL-1; Galaptin; Galbp; galectin 1; galectin 1; LGALS1; Galectin1; galectin-1; galectin-1-like protein; GBP; HBL; H-Galectin-1; HLBP14; HPL; L14; L-14.5; Lactose-binding lectin 1; Lect14; Lectin galactoside-binding soluble 1; lectin, galactose binding, soluble 1; lectin, galactoside binding soluble 1; lectin, galactoside-binding, soluble, 1; lectin, galactoside-binding, soluble, 1 (galectin 1); Lgals1; LGAS1; OPG-1; pm12; Putative MAPK-activating protein PM12; RL 14.5; similar to galactose-binding lectin; S-Lac lectin 1; unnamed protein product
Common Name Galectin 1
Gene Symbol LGALS1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LKPGECLRVRGEVAPDAKSFVLNLGKDSNNLCLHFNPRFNAHGDANTIVCNSKDGGAWGTEQREAVFPFQPGSVAEVCITFDQANLTVKLPDGYEFKFPNRLNLEAINYMAADGDF
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.