Learn More
R&D Systems™ Human Galectin-8 (NP_963837.1) VersaClone cDNA
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Specifications
Specifications
| Gene | Galectin-8 |
| Accession Number | NP_963837.1 |
| Gene ID (Entrez) | 3964 |
| Gene Alias | Gal-8, GAL8, galectin 8, galectin-8, Galectin8, galectin-8 g, lectin, galactoside-binding, soluble, 8, LGALS8, PCTA1, PCTA-1 Po66 carbohydrate-binding protein, Po66 carbohydrate binding protein, Po66-CBP, Prostate carcinoma tumor antigen 1 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |