missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GATA-3 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC26052G-100UG
This item is not returnable.
View return policy
Description
GATA-3 Monoclonal specifically detects GATA-3 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| GATA-3 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| 131320, GATA binding protein 3, GATA3, GATA-binding factor 3, GATA-binding protein 3, HDRMGC5199, MGC2346, MGC5445, trans-acting T-cell-specific transcription factor GATA-3 | |
| E. coli-derived recombinant human GATA-3, Pro135-Ser258, Accession # P23771 | |
| 100 μg | |
| Primary | |
| Detects human GATA-3 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG2b |
| Flow Cytometry | |
| 634919 | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| P23771 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2625 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction