Learn More
R&D Systems™ Human GDNF Quantikine ELISA Kit
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Description
The Human GDNF Quantikine ELISA Kit quantifies human GDNF in cell culture supernates (50 μL), cell lysates (50 μL), tissue lysates (50 μL), serum (50 μL), edta plasma (50 μL), heparin plasma (50 μL).
Specifications
Specifications
| Assay Range | 4.69 to 300 pg/mL |
| Assay Sensitivity | 0.696 pg/mL |
| Sample Type | Cell Culture Supernatant, Cell Lysates, Plasma (EDTA, Heparin), Serum, Tissue Lysates |
| Specificity | Natural and Recombinant Mouse ELA2 |
| Gene ID (Entrez) | 2668 |
| Host Species | Human |
| Quantity | 1 Kit |
| Sample Volume | Cell Culture Supernates (50 μL),Cell Lysates (50 μL),Tissue Lysates (50 μL),Serum (50 μL),EDTA Plasma (50 μL),Heparin Plasma (50 μL) |
| Storage Requirements | Store the unopened product at -20° to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Total Assay Time | 4 hr. |