missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Human GHITM ELISA Kit (Chemiluminescence)
Shop All Bio Techne Products
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Specifications
Specifications
| Assay Range | 62.50 to 4000 pg/mL |
| Assay Sensitivity | 37.50 pg/mL |
| Product Type | ELISA Kit (Chemiluminescence) |
| Sample Type | Serum, Plasma, Biological Fluids |
| Specificity | This kit recognizes Human GHITM in samples. No significant cross-reactivity or interference between Human GHITM and analogues was observed. |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 27069 |
| Gene Symbol | GHITM |
| Quantity | 1 Kit |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction