missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GloLIVE TRA-1-60(R) NorthernLights™ NL557-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems NLLC4770R
This item is not returnable.
View return policy
Description
TRA-1-60(R) Monoclonal specifically detects TRA-1-60(R) in Human samples. It is validated for Immunocytochemistry.
Specifications
| TRA-1-60(R) | |
| Monoclonal | |
| NorthernLights 557 (NL557) | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| PODXL | |
| 2102Ep human embryonal carcinoma cell line. | |
| 0.5 mL | |
| Primary | |
| Human | |
| IgM |
| Immunocytochemistry | |
| TRA-1-60 | |
| Immunocytochemistry | |
| GCTM, PCLP, POD XL, PODXL, RA160, TRA160(R), tra61 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Detects human TRA-1-60(R). | |
| Store the unopened product at 2 - 8degrees C. Do not use past expiration date. Protect from light. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction