missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GLP-1R Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB28141-SP
This item is not returnable.
View return policy
Description
GLP-1R Monoclonal specifically detects GLP-1R in Human samples. It is validated for Immunohistochemistry.
Specifications
| GLP-1R | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| P43220 | |
| GLP1R | |
| Baf/3 cell line transfected with human GLP-1R Accession # P43220 | |
| 25 μg | |
| Primary | |
| Detects human GLP-1R. Stains human GLP-1R-transfected Baf/3 cells but not irrelevant transfectants. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 419208 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| GLP-1 receptor, GLP1R, GLP-1R, GLP-1-R, glucagon-like peptide 1 receptor, MGC138331 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2740 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction