missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GLP-1R Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB28141
This item is not returnable.
View return policy
Description
GLP-1R Monoclonal specifically detects GLP-1R in Human samples. It is validated for Immunohistochemistry.
Specifications
| GLP-1R | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| P43220 | |
| GLP1R | |
| Baf/3 cell line transfected with human GLP-1R Accession # P43220 | |
| 100 μg | |
| Primary | |
| Detects human GLP-1R. Stains human GLP-1R-transfected Baf/3 cells but not irrelevant transfectants. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 419208 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| GLP-1 receptor, GLP1R, GLP-1R, GLP-1-R, glucagon-like peptide 1 receptor, MGC138331 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2740 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction