Learn More
Human GLP-2R Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems FAB4285G-100UG
Product Code. 18782498
  • € 592.00 / 100µg
Description
GLP-2R Monoclonal specifically detects GLP-2R in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GLP-2R
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
O95838
GLP2R
HEK293 human embryonic kidney cell line transfected with human GLP-2R Met1-Ile553 Accession # O95838
RUO
9340
Human
IgG2a
Flow Cytometry
413801
Alexa Fluor 488
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
GLP-2 receptor, GLP2R, GLP-2R, GLP-2-R, glucagon-like peptide 2 receptor
Mouse
100 μg
Primary
Detects human GLP-2R. Stains human GLP-2R transfectants but not irrelevant transfectants.
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
For Research Use Only (RUO)