Learn More
Human GLP-2R Alexa Fluor 488-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems FAB4285G-100UG
This item is not returnable.
View return policy
Description
GLP-2R Monoclonal specifically detects GLP-2R in Human samples. It is validated for Flow Cytometry.
Specifications
| GLP-2R | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| O95838 | |
| GLP2R | |
| HEK293 human embryonic kidney cell line transfected with human GLP-2R Met1-Ile553 Accession # O95838 | |
| RUO | |
| 9340 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 413801 | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| GLP-2 receptor, GLP2R, GLP-2R, GLP-2-R, glucagon-like peptide 2 receptor | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human GLP-2R. Stains human GLP-2R transfectants but not irrelevant transfectants. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Product Title
For Research Use Only (RUO)