missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GLP-2R Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB4285R-100UG
This item is not returnable.
View return policy
Description
GLP-2R Monoclonal specifically detects GLP-2R in Human samples. It is validated for Flow Cytometry.
Specifications
| GLP-2R | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| O95838 | |
| GLP2R | |
| HEK293 human embryonic kidney cell line transfected with human GLP-2R Met1-Ile553 Accession # O95838 | |
| RUO | |
| 9340 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 413801 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| GLP-2 receptor, GLP2R, GLP-2R, GLP-2-R, glucagon-like peptide 2 receptor | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human GLP-2R. Stains human GLP-2R transfectants but not irrelevant transfectants. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction