missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GluR6/GRIK2 Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB9610G-100UG
This item is not returnable.
View return policy
Description
KLHL24 Monoclonal antibody specifically detects KLHL24 in Human samples. It is validated for Flow Cytometry
Specifications
| KLHL24 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| DRE1, EAA4, GLR6, GluK2, GLUK6, Kainate Receptor Interacting Protein For GluR6, Kainate Receptor-Interacting Protein For GluR6, Kelch-Like 24, Kelch-Like 24 (Drosophila), Kelch-Like Family Member 24, Kelch-Like Protein 24, KLHL24, KRIP6, MRT6, Protein DRE1 | |
| Chinese Hamster Ovary cell line CHO-derived human GluR6/GRIK2 protein, Thr32-Asn549, Accession # Q13002 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 818235 | |
| Flow Cytometry | |
| Q13002 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 54800 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction