missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GluR7/GRIK3 Alexa Fluor™ 750-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems FAB113281S-100UG
This item is not returnable.
View return policy
Description
GluR7/GRIK3 Monoclonal antibody specifically detects GluR7/GRIK3 in Human samples. It is validated for Flow Cytometry
Specifications
| GluR7/GRIK3 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| Q13003 | |
| Rabbit | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2899 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2°C to 8°C as supplied. | |
| IgG |
| Flow Cytometry | |
| 2914A | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| EAA5, Excitatory amino acid receptor 5, GLR7, gluR-7, GluR7, GluR7a, GLUR7dJ1090M5.1 (glutamate receptor, ionotropic, kainate 3 (GLUR7)), Glutamate receptor 7, glutamate receptor, ionotropic kainate 3, glutamate receptor, ionotropic, kainate 3 | |
| Human embryonic kidney cell, HEK293-derived human GluR7/GRIK3 , Gly249-Met1, Accession # Q13003 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction