missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Glut4 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB86541R-025
This item is not returnable.
View return policy
Description
Glut4 Monoclonal specifically detects Glut4 in Human samples. It is validated for Flow Cytometry.
Specifications
| Glut4 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Supplied in a saline solution containing BSA and Sodium Azide. | |
| GLUT-4, GLUT4Glucose transporter type 4, insulin-responsive, insulin-responsive glucose transporter type 4, SLC2A4, solute carrier family 2 (facilitated glucose transporter), member 4, solute carrier family 2, facilitated glucose transporter member 4 | |
| Mouse myeloma cell line NS0-derived recombinant human Glut4 Met1-Asp509 Accession # P14672 | |
| 25 Tests | |
| Primary | |
| Detects human Glut4 in direct ELISAs. Stains human Glut4 transfectants but not irrelevant transfectants in flow cytometry. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
| Flow Cytometry | |
| 925932 | |
| Flow Cytometry 5 uL/10^6 cells | |
| P14672 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6517 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction