missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Glycine N-methyltransferase/GNMT Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF6526-SP
This item is not returnable.
View return policy
Description
Glycine N-Methyltransferase/GNMT Polyclonal specifically detects Glycine N-Methyltransferase/GNMT in Human samples. It is validated for Western Blot.
Specifications
| Glycine N-Methyltransferase/GNMT | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 2.1.1.20, glycine N-methyltransferase, Glycine NMethyltransferase, GNMT | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 27232 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q14749 | |
| GNMT | |
| E. coli-derived recombinant human Glycine N-methyltransferase/GNMT Met1-Asp295 Accession # Q14749 | |
| 25 μg | |
| Primary | |
| Detects human Glycine N-methyltransferase/GNMT in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction