missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Glycoprotein V/CD42d Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF4249-SP
This item is not returnable.
View return policy
Description
Glycoprotein V/CD42d Polyclonal specifically detects Glycoprotein V/CD42d in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| Glycoprotein V/CD42d | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| platelet glycoprotein V, CD42d, glycoprotein V (platelet), GP5, GPV | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 2814 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| P40197 | |
| GP5 | |
| Mouse myeloma cell line NS0-derived recombinant human Glycoprotein V/CD42d Gln17-Gly523 Accession # P40197 | |
| 25 μg | |
| Primary | |
| Detects human Glycoprotein V/CD42d in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human GPVI is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction