missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Glypican 1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems AF4519-SP
This item is not returnable.
View return policy
Description
Glypican 1 Polyclonal specifically detects Glypican 1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Specifications
| Glypican 1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FLJ38078, glypican, glypican 1, glypican proteoglycan 1, glypican-1, GPC1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2817 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL, CyTOF-ready | |
| P35052 | |
| GPC1 | |
| Mouse myeloma cell line NS0-derived recombinant human Glypican 1 Asp24-Ser530 Accession # P35052 | |
| 25 μg | |
| Primary | |
| Detects human Glypican 1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 15% cross-reactivity with recombinant mouse Glypican 1 is observed and less than 1% cross-reactivity with recombinant human (rh) Glypican 5 and rhGlypican 6 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction