missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Glypican 3 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB2119T-100UG
This item is not returnable.
View return policy
Description
Glypican 3 Monoclonal specifically detects Glypican 3 in Human samples. It is validated for Flow Cytometry.
Specifications
| Glypican 3 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P51654.1 | |
| GPC3 | |
| Mouse myeloma cell line NS0-derived recombinant human Glypican 3 Gln25-Val558 Accession # P51654.1 | |
| RUO | |
| 2719 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 307801 | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| DGSX, glypican 3, glypican proteoglycan 3, glypican-3, GPC3, GTR2-2, heparan sulphate proteoglycan, Intestinal protein OCI-5, MXR7, OCI5, OCI-5, secreted glypican-3, SGB, SGBS, SGBS1SDYS | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human Glypican 3 in ELISAs and Western blots. Does not cross-react with recombinant human (rh) Glypican-2, rhGlypican-5, or rhGlypican-6. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only (RUO)
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido