missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GM-CSF Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC615R-100UG
This item is not returnable.
View return policy
Description
GM-CSF Monoclonal specifically detects GM-CSF in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| GM-CSF | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| P04141 | |
| CSF2 | |
| E. coli-derived recombinant human GM-CSF Ala18-Glu144 Accession # P04141 | |
| RUO | |
| 1437 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 6804 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, CSF2, CSF-2, GMCSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, Molgramostim, molgramostin, sargramostim | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human GM-CSF in ELISAs. In ELISAs, no cross-reactivity with recombinant mouse GM-CSF, recombinant rat GM-CSF, recombinant human (rh) M-CSF, rhG-CSF, rhIL-5-R alpha , or rhIL-3 R alpha is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction