missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GM-CSF Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 6 publications
Brand: R&D Systems AF-215-NA
This item is not returnable.
View return policy
Description
GM-CSF Polyclonal specifically detects GM-CSF in Human samples. It is validated for Western Blot, Neutralization.
Specifications
| GM-CSF | |
| Polyclonal | |
| Unconjugated | |
| P04141 | |
| CSF2 | |
| E. coli-derived recombinant human GM-CSF Ala18-Glu144 Accession # P04141 | |
| 100 μg | |
| Primary | |
| Detects human GM-CSF in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse GM-CSF is observed. Neutralizes the biological activity of both recombinant human GM-CSF and natural human GM-CSF. | |
| Human | |
| IgG |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 0.08-0.16 ug/mL | |
| colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, CSF2, CSF-2, GMCSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, Molgramostim, molgramostin, sargramostim | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1437 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction