missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human gp130 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB228G-100UG
This item is not returnable.
View return policy
Description
gp130/CD130 Monoclonal specifically detects gp130/CD130 in Human samples. It is validated for Flow Cytometry.
Specifications
| gp130/CD130 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| CD130, CD130 antigen, CDw130, DKFZp564F053, gp130, gp130 of the rheumatoid arthritis antigenic peptide-bearing soluble form, IL-6 receptor subunit beta, IL-6R subunit beta, IL-6RB, IL-6R-beta, IL6ST, interleukin 6 signal transducer (gp130, oncostatin M receptor), interleukin receptor beta chain, interleukin-6 receptor subunit beta, Interleukin-6 signal transducer, Membrane glycoprotein 130, membrane glycoprotein gp130, Oncostatin-M receptor subunit alpha | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human gp130 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) IL-2 R, rhIL-4 R, or rhIL-6 R is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
| Flow Cytometry | |
| 28126 | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| IL6ST | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human gp130 extracellular domain | |
| RUO | |
| 3572 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction