missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GPR158 Alexa Fluor™ 594-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems FAB10286T-100UG
This item is not returnable.
View return policy
Description
GPR158 Monoclonal antibody specifically detects GPR158 in Human samples. It is validated for Flow Cytometry
Specifications
| GPR158 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| G protein-coupled receptor 158, KIAA1136, KIAA1136FLJ37801, probable G-protein coupled receptor 158, RP11-59G22.1 | |
| Human embryonic kidney cell HEK293-derived human GPR158 protein, Ala24-Gln411, Accession # Q5T848 | |
| 100 μg | |
| Primary | |
| Reconstitution default not known | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2°C to 8°C as supplied. | |
| IgG2b |
| Flow Cytometry | |
| 1027604 | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q5T848 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 57512 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction