missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GPR34 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC4617V-100UG
This item is not returnable.
View return policy
Description
GPR34 Monoclonal specifically detects GPR34 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| GPR34 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q9UPC5 | |
| GPR34 | |
| NS0 mouse myeloma cell line transfected with human GPR34 Accession # Q9UPC5 | |
| RUO | |
| 2857 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 419859 | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| G protein-coupled receptor 34, probable G-protein coupled receptor 34 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Stains human GPR34 transfectants but not irrelevant transfectants. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction