missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GPR37 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC4450U-100UG
This item is not returnable.
View return policy
Description
GPR37 Monoclonal specifically detects GPR37 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| GPR37 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| O15354 | |
| GPR37 | |
| NS0 mouse myeloma cell line transfected with human GPR37 Ala27-Cys613 Accession # O15354 | |
| RUO | |
| 2861 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 436923 | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| EDNRBL, Endothelin B receptor-like protein 1, ETBR-LP-1, G protein-coupled receptor 37 (endothelin receptor type B-like), hET(B)R-LP, PAELR, PAELRETBR-LP-1, Parkin-associated endothelin receptor-like receptor, probable G-protein coupled receptor 37 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human GPR37. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction