missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GPR83 Alexa Fluor™ 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB10943N-100UG
This item is not returnable.
View return policy
Description
GPR83 Monoclonal antibody specifically detects GPR83 in Human samples. It is validated for Flow Cytometry
Specifications
| GPR83 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide | |
| G protein-coupled receptor 72, G protein-coupled receptor 83, GIR, GPR72, GPR72glucocorticoid induced receptor, G-protein coupled receptor 72, KIAA1540, probable G-protein coupled receptor 83, RP39, RP82 | |
| NS0 mouse myeloma cell line transfected with human GPR83, Accession # Q9NYM4 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 566133 | |
| Flow Cytometry | |
| Q9NYM4 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 10888 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction