missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GPRC5A Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC5239S-100UG
This item is not returnable.
View return policy
Description
GPRC5A/RAI3 Monoclonal specifically detects GPRC5A/RAI3 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| GPRC5A/RAI3 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q8NFJ5 | |
| GPRC5A | |
| NS0 mouse myeloma cell line transfected with human GPRC5A Met1-Ser357 Accession # Q8NFJ5 | |
| RUO | |
| 9052 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 481906 | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| G protein-coupled receptor, family C, group 5, member A, GPCR5A, Orphan G-protein-coupling receptor PEIG-1, RAI3, RAI3G-protein coupled receptor family C group 5 member A, RAIG1, RAIG1RAIG-1, retinoic acid induced 3, retinoic acid responsive, Retinoic acid-induced gene 1 protein, retinoic acid-induced protein 3 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Stains human GPRC5A transfectants but not irrelevant transfectants. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction