missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GPRC5C Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB6594S-100UG
This item is not returnable.
View return policy
Description
GPRC5C Monoclonal specifically detects GPRC5C in Human samples. It is validated for Flow Cytometry.
Specifications
| GPRC5C | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q9NQ84 | |
| GPRC5C | |
| NS0 mouse myeloma cell line transfected with recombinant human GPRC5C Accession # Q9NQ84 | |
| RUO | |
| 55890 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 577315 | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| G protein-coupled receptor, family C, group 5, member C, MGC131820, RAIG3, RAIG-3G-protein coupled receptor family C group 5 member C, RAIG3orphan G-protein coupled receptor, retinoic acid responsive gene protein, Retinoic acid-induced gene 3 protein | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human GPRC5C in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction