missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Granzyme A Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF2905
This item is not returnable.
View return policy
Description
Granzyme A Polyclonal specifically detects Granzyme A in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry.
Specifications
| Granzyme A | |
| Polyclonal | |
| Unconjugated | |
| P12544 | |
| GZMA | |
| Mouse myeloma cell line NS0-derived recombinant human Granzyme A Cys26-Val262 Accession # P12544 | |
| 100 μg | |
| Primary | |
| Detects human Granzyme A in direct ELISAs. | |
| Human | |
| IgG |
| Western Blot, ELISA, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.25 ug/mL, ELISA, Immunocytochemistry 5-15 ug/mL | |
| CTL tryptase, CTLA3, CTLA3Granzyme A (Cytotoxic T-lymphocyte-associated serine esterase-3; Hanukah factorserine protease), Cytotoxic T-lymphocyte proteinase 1, EC 3.4.21, fragmentin-1, granzyme A, granzyme A (granzyme 1, cytotoxic T-lymphocyte-associated serine esterase 3), Granzyme-1, GZMA, H factor, Hanukkah factor, HF, HFSP, HFSPEC 3.4.21.78 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3001 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction