Learn More
Human Growth Hormone Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB1067-500
 View more versions of this product
Product Code. 18782689
  • ONLINE EXCLUSIVE PRICE
    € 2020.00
    € 1909.95 / 500µg
    Save € 110.05
    5% Off
Description
Growth Hormone Monoclonal specifically detects Growth Hormone in Human samples. It is validated for Western Blot, Neutralization.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Growth Hormone
Monoclonal
LYOPH
Western Blot 2 ug/mL, Neutralization 2-8 ug/mL
CAA23779
GH1
E. coli-derived recombinant human Growth Hormone Phe27-Phe217 Accession # CAA23779
500 μg
Primary
Detects human Growth Hormone (GH) in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant rat GH is observed.
Human
Supernatant
Western Blot, Neutralization
178902
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
GH1, GHB5, GHIGHD1B, GHN, GH-N, GHNGrowth hormone, GH-Nsomatotropin, growth hormone 1Pituitary growth hormone, hGH-N, IGHD1A, IGHD2, Somatotropin
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
2688
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2b