missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GSK-3 alpha Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB2986-SP
This item is not returnable.
View return policy
Description
GSK-3 alpha Monoclonal specifically detects GSK-3 alpha in Human samples. It is validated for Immunocytochemistry.
Specifications
| GSK-3 alpha | |
| Monoclonal | |
| LYOPH | |
| Immunocytochemistry 8-25 ug/mL | |
| P49840.2 | |
| GSK3A | |
| E. coli-derived recombinant human GSK-3 alpha Gly90-Ser483 Accession # P49840.2 | |
| 25 μg | |
| Primary | |
| Detects human GSK-3 alpha . In direct ELISAs, this antibody does not detect recombinant human GSK-3 beta . | |
| Human | |
| Purified |
| Immunocytochemistry | |
| 288901 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DKFZp686D0638, EC 2.7.11, EC 2.7.11.26, glycogen synthase kinase 3 alpha, glycogen synthase kinase-3 alpha, GSK3 alpha, GSK-3 alpha, GSK3A | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2931 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction