Learn More
Novus Biologicals™ Human Heart Atrium Whole Tissue Lysate (Adult Whole Arrhythmia)
Shop All Bio Techne ProductsDescription
The Human Heart Atrium Whole Tissue Lysate (Adult Whole Arrhythmia) is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation.
Specifications
Specifications
| For Use With (Application) | Western Blot |
| Tissue | Heart |
| Quantity | 1 mg |
| Storage Buffer | HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors |
| Test Specificity | Total Protein - Arrhythmia, infarct: Heart Atrium, right |
| Cellular Fraction | Whole |
| Tissue Condition | Arrhythmia |
| Cross Reactivity | Human |
| Host Species | Human |
| Concentration | 5 mg/mL |
| Show More |