missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Hemoglobin Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems G-134-C
This item is not returnable.
View return policy
Description
Hemoglobin Polyclonal specifically detects Hemoglobin in Human samples. It is validated for .
Specifications
| Hemoglobin | |
| LYOPH | |
| Hidden | |
| Affinity Purified | |
| RUO | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Store the unopened product at 2 - 8degrees C. |
| Polyclonal | |
| Unconjugated | |
| Goat | |
| 0.1 mg | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction