Learn More
Human Heparan Sulfate 6-O-Sulfotransferase 2/HS6ST2 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand:  R&D Systems AF2710
 View more versions of this product
Product Code. 18029411
  • € 501.00 / 100µg
Description
Heparan Sulfate 6-O-Sulfotransferase 2/HS6ST2 Polyclonal specifically detects Heparan Sulfate 6-O-Sulfotransferase 2/HS6ST2 in Human samples. It is validated for Western Blot, Immunoprecipitation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Heparan Sulfate 6-O-Sulfotransferase 2/HS6ST2
Polyclonal
Unconjugated
Q96MM7
HS6ST2
Mouse myeloma cell line NS0-derived recombinant human Heparan Sulfate 6-O-Sulfotransferase 2/HS6ST2 Gln179-Arg605 Accession # Q96MM7
100 μg
Primary
Detects human Heparan Sulfate 6-O-Sulfotransferase 2/HS6ST2 in direct ELISAs and Western blots.
Human
IgG
Western Blot, Immunoprecipitation
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
EC-1, heparan sulfate 6-O-sulfotransferase 2
Goat
Affinity Purified
RUO
90161
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.