missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human HES-4 Biotinylated Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems BAF3600
This item is not returnable.
View return policy
Description
HES-4 Polyclonal specifically detects HES-4 in Human samples. It is validated for Western Blot.
Specifications
| HES-4 | |
| Polyclonal | |
| Biotin | |
| Q9HCC6 | |
| HES4 | |
| E. coli-derived recombinant human HES-4 Val94-Arg221 Accession # Q9HCC6 | |
| 50 μg | |
| Primary | |
| Detects human HES-4 in Western blots. In Western blots, less than 1% cross-reactivity with recombinant human (rh) HES-1, rhHES-2, rhHES-5, and rhHES-7 is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| bHLH factor Hes4, BHLHB42, bHLHb42transcription factor HES-4, Class B basic helix-loop-helix protein 42, Hairy and enhancer of split 4, hairy and enhancer of split 4 (Drosophila), HES4, hHES4 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 57801 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction