missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Hexokinase 1/2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB8179-SP
This item is not returnable.
View return policy
Description
Hexokinase 1/2 Monoclonal specifically detects Hexokinase 1/2 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.
Specifications
| Hexokinase 1/2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Detects human Hexokinase 2 in direct ELISAs. Detects human Hexokinase 1 and human Hexokinase 2 in Western blots. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 927312 | |
| Western Blot 0.2 ug/mL, Simple Western 5 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| P52789 | |
| E. coli-derived recombinant human Hexokinase 2. Phe11-Arg917 Accession # P52789 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction