missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human HIC5/TGFB1I1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF5626-SP
This item is not returnable.
View return policy
Description
HIC5/TGFB1I1 Polyclonal specifically detects HIC5/TGFB1I1 in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| HIC5/TGFB1I1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Androgen receptor coactivator 55 kDa protein, androgen receptor coactivator ARA55, Androgen receptor-associated protein of 55 kDa, ARA55, ARA55Hydrogen peroxide-inducible clone 5 protein, HIC5, HIC-5, TGFB1I1, transforming growth factor beta 1 induced transcript 1, transforming growth factor beta-1-induced transcript 1 protein, TSC-5 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7041 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| O43294 | |
| TGFB1I1 | |
| E. coli-derived recombinant human HIC5 Val184-Gly288 Accession # O43294 | |
| 25 μg | |
| Primary | |
| Detects endogenous human HIC5 in Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction