missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human HIKESHI (aa 115-196) Control Fragment Recombinant Protein

Product Code. 30204493
missing translation for 'orderingAttributeHoverText'
Quantity:
100 μL
missing translation for 'unitSize'
100µL
This item is not returnable. View return policy

Product Code. 30204493

missing translation for 'mfr': Invitrogen™ RP96567

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (95%), Rat (95%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57139 (PA5-57139. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Required for organization and/or function of the secretory apparatus in Clara cells in lung.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q53FT3
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 51501
Name Human HIKESHI (aa 115-196) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 0610007P06Rik; 1110002N09Rik; C11orf73; C1H11orf73; C21H11orf73; C29H11orf73; chromosome unknown open reading frame, human C11orf73; heat shock protein nuclear import factor hikeshi; Hikeshi; Hikeshi, heat shock protein nuclear import factor; HLD13; HSPC138; HSPC179; HSPC248; hypothetical protein LOC504867; hypothetical protein LOC550239; l(7)6 Rn; l7Rn6; lethal gene on chromosome 7 Rinchik 6 protein; lethal, Chr 7, Rinchik 6; LOC100762248; OPI10; protein Hikeshi; Protein of unknown function DUF775 containing protein; similar to hypothetical protein HSPC138; uncharacterized protein C11orf73 homolog; Uncharacterized protein C11orf73-like; unknown; zgc:110091
Common Name HIKESHI
Gene Symbol HIKESHI
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence VELLDSMAQQTPVGNAAVSSVDSFTQFTQKMLDNFYNFASSFAVSQAQMTPSPSEMFIPANVVLKWYENFQRRLAQNPLFWK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.