Learn More
Human Histamine H1 R Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB4726-SP
Product Code. 18747148
  • € 209.00 / 25µg
Description
Histamine H1R Monoclonal specifically detects Histamine H1R in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Histamine H1R
Monoclonal
LYOPH
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
AAH60802
HRH1
NS0 mouse myeloma cell line transfected with human Histamine H1 R Met1-Ser487 Accession # AAH60802
25 μg
Primary
Detects human Histamine H1 R. Stains human Histamine H1 R transfectants but not irrelevant transfectants.
Human
Purified
Flow Cytometry, CyTOF
480054
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
H1R, H1-R, hisH1, Histamine H1R, histamine receptor H1, HRH1, subclass H1
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
3269
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.