missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human hnRNP H2 (aa 45-161) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (99%), Rat (99%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53513 (PA5-53513. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
This gene belongs to the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene has three repeats of quasi-RRM domains that binds to RNAs. It is very similar to the family member HNRPH1. This gene is thought to be involved in Fabray disease and X-linked agammaglobulinemia phenotype. Alternative splicing results in multiple transcript variants encoding the same protein. [provided by RefSeq].
Specifications
Specifications
| Accession Number | P55795 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 3188 |
| Name | Human hnRNP H2 (aa 45-161) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | DXHXS1271E; Ftp3; Ftp-3; H'; Heterogeneous nuclear ribonucleoprotein H'; heterogeneous nuclear ribonucleoprotein H2; heterogeneous nuclear ribonucleoprotein H2 (H'); Heterogeneous nuclear ribonucleoprotein H2, N-terminally processed; heterogeneous nuclear ribonucleoprotein H-prime; HNRNP; hnRNP H'; hnRNP H2; hnRNPH'; Hnrnph2; HNRPH'; HNRPH2 |
| Common Name | hnRNP H2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction