missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human HOXC8 (aa 28-149) Control Fragment Recombinant Protein

Product Code. 30196555
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Product Code. Quantity unitSize
30196555 100 μL 100µL
1 options
This item is not returnable. View return policy

Product Code. 30196555

Brand: Invitrogen™ RP102308

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-83043 (PA5-83043. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, which are located on different chromosomes and consist of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXC genes located in a cluster on chromosome 12. The product of this gene may play a role in the regulation of cartilage differentiation. It could also be involved in chondrodysplasias or other cartilage disorders.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P31273
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 3224
Name Human HOXC8 (aa 28-149) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias D130011F21Rik; homeo box 3 A; homeo box C8; homeobox C8; Homeobox gene C8; homeobox protein Hox-3.1; Homeobox protein Hox-3 A; homeobox protein Hox-C8; Homeobox protein M31; homeobox protein R4; HO x 3; Hox-3.1; Hox-3.1, mouse, homolog of; HO x 3 A; Ho x 3r4; HOXC8; Hoxc-8
Common Name HOXC8
Gene Symbol HOXC8
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence FPQSVGRSHALVYGPGGSAPGFQHASHHVQDFFHHGTSGISNSGYQQNPCSLSCHGDASKFYGYEALPRQSLYGAQQEASVVQYPDCKSSANTNSSEGQGHLNQNSSPSLMFPWMRPHAPGR
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.