missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human HSCB (aa 160-232) Control Fragment Recombinant Protein

Product Code. 30212804
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30212804

Brand: Invitrogen™ RP92350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (81%), Rat (81%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53802 (PA5-53802. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

May act as a co-chaperone in iron-sulfur cluster assembly in mitochondria.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8IWL3
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 150274
Name Human HSCB (aa 160-232) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AI325508; AW049829; C-HSC20; dJ366L4.2; DnaJ (Hsp40) homolog, subfamily C, member 20; DnaJ homolog subfamily C member 20; DNAJC20; HSC20; HSCB; HscB iron-sulfur cluster co-chaperone homolog; HscB iron-sulfur cluster co-chaperone homolog (E. coli); HscB mitochondrial iron-sulfur cluster cochaperone; HscB mitochondrial iron-sulfur cluster co-chaperone; iron-sulfur cluster co-chaperone protein HscB; Iron-sulfur cluster co-chaperone protein HscB, cytoplasmic; Iron-sulfur cluster co-chaperone protein HscB, mitochondrial; JAC1; J-type co-chaperone HSC20; RGD1311005
Common Name HSCB
Gene Symbol HSCB
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence QFLIEIMEINEKLAEAESEAAMKEIESIVKAKQKEFTDNVSSAFEQDDFEEAKEILTKMRYFSNIEEKIKLKK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.