missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human HTR3A (aa 343-435) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (81%), Rat (81%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
The product of this gene belongs to the ligand-gated ion channel receptor superfamily. This gene encodes subunit A of the type 3 receptor for 5-hydroxytryptamine, a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. This receptor causes fast, depolarizing responses in neurons after activation. It appears that the heteromeric combination of A and B subunits is necessary to provide the full functional features of this receptor, since either subunit alone results in receptors with very low conductance and response amplitude. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
Specifications
| Accession Number | P46098 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 3359 |
| Name | Human HTR3A (aa 343-435) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 5 HT3; 5-HT3; 5-HT-3; 5-HT3 receptor; 5 HT3 serotonin receptor; 5-HT3A; 5-HT3-A; 5 HT3R; 5-HT3R; 5-hydroxytryptamine (serotonin) receptor 3 A; 5-hydroxytryptamine (serotonin) receptor 3 A, ionotropic; 5-hydroxytryptamine receptor 3; 5-hydroxytryptamine receptor 3 A; Htr3; Htr3a; serotonin receptor; Serotonin receptor 3 A; serotonin-gated ion channel receptor; truncated receptor |
| Common Name | HTR3A |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction